Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-L-phenylalanine, 98%, Thermo Scientific™

Catalog No. AAH26905MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH26905MD 250 mg
AAH2690503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH26905MD Supplier Thermo Scientific Chemicals Supplier No. H26905MD

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Used in absorption liquids on a technical scale for removing acid components such as carbon dioxide and hydrogen sulfide from gases. 4-Methyl-L-phenylalanine are used in buffers to maintain the pH of aqueous liquids at a constant level.

Solubility
Sparingly soluble (0.014 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1991-87-3
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.22
MDL Number MFCD00038575
InChI Key DQLHSFUMICQIMB-VIFPVBQESA-N
Synonym 4-methyl-l-phenylalanine, l-4-methylphenylalanine, 4-methylphenylalanine, h-phe 4-me-oh, 2s-2-amino-3-4-methylphenyl propanoic acid, phenylalanine, 4-methyl, s-2-amino-3-p-tolyl propanoic acid, l-4-me-phe-oh, l-4-methylphe, 4-methyl-l-phe-oh
PubChem CID 151513
IUPAC Name (2S)-2-amino-3-(4-methylphenyl)propanoic acid
SMILES CC1=CC=C(C[C@H](N)C(O)=O)C=C1

Specifications

Solubility Information Sparingly soluble (0.014g/L) (25°C).
Quantity 250 mg
Formula Weight 179.22
Percent Purity 98%
Chemical Name or Material 4-Methyl-L-phenylalanine
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.