Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-n-octanoic Acid 98.0+%, TCI America™
SDP

Catalog No. M20485G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M20485G Supplier TCI America Supplier No. M20485G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 54947-74-9
Molecular Formula C9H17O2
Molecular Weight (g/mol) 157.23
MDL Number MFCD00051938
InChI Key LEGGANXCVQPIAI-MRVPVSSYSA-M
Synonym octanoic acid, 4-methyl, 4-methyl-octanoic acid, 4-methylcaprylic acid, +/--4-methyloctanoic acid, 4-methyl-n-octanoic acid, 4-methyl octanoic acid, octanoic acid, 4-methyl-, r, fema no. 3575, acmc-20mssw, acmc-20mssx
PubChem CID 62089
IUPAC Name (4R)-4-methyloctanoate
SMILES CCCC[C@@H](C)CCC([O-])=O

Specifications

Color Yellow
Boiling Point 143°C
Quantity 5 g
UN Number 3265
Formula Weight 158.24
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 4-Methyl-n-octanoic Acid
EINECSNumber (2)-0608
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.