Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-N-tosylbenzenesulfonamide 98.0+%, TCI America™
SDP

Catalog No. M29751G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M29751G 1 g
M29755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M29751G Supplier TCI America Supplier No. M29751G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3695-00-9
Molecular Formula C14H15NO4S2
Molecular Weight (g/mol) 325.397
MDL Number MFCD00014917
InChI Key LHWZLUXODWUHLZ-UHFFFAOYSA-N
Synonym bis p-tolylsulfonyl amine, di-p-toluenesulfonamide, ditosylamine, di-p-toluenesulfonimide, bis 4-methylbenzenesulfon amine, benzenesulfonamide, 4-methyl-n-4-methylphenyl sulfonyl, 4-methyl-n-tosylbenzenesulfonamide, 4-methyl-n-4-methylphenyl sulfonyl benzenesulfonamide
PubChem CID 77279
IUPAC Name 4-methyl-N-(4-methylphenyl)sulfonylbenzenesulfonamide
SMILES CC1=CC=C(C=C1)S(=O)(=O)NS(=O)(=O)C2=CC=C(C=C2)C

Specifications

Melting Point 173°C
Color White
Boiling Point 265°C
Quantity 1 g
Formula Weight 325.40
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methyl-N-tosylbenzenesulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.