Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-N1-(3-phenylpropyl)-1,2-phenylenediamine 98.0+%, TCI America™
SDP

Catalog No. M2734100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2734100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M2734100MG Supplier TCI America Supplier No. M2734100MG

Chemical Identifiers

CAS 749886-87-1
Molecular Formula C16H20N2
Molecular Weight (g/mol) 240.35
MDL Number MFCD09753595
InChI Key YMFNPBSZFWXMAD-UHFFFAOYSA-N
Synonym JSH-23
PubChem CID 16760588
IUPAC Name 4-methyl-N1-(3-phenylpropyl)benzene-1,2-diamine
SMILES CC1=CC(N)=C(NCCCC2=CC=CC=C2)C=C1

Specifications

Melting Point 103°C
Quantity 100 mg
Formula Weight 240.35
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methyl-N1-(3-phenylpropyl)-1,2-phenylenediamine
TSCA No
Recommended Storage Freezer (-20 °C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.