Learn More
Cayman Chemical 4-Methylumbelliferyl Palmitate, 17695-48-6, 1 g
SDP
Supplier:  Cayman Chemical 160891
Encompass_Preferred
A fluorogenic substrate for LAL; is cleaved by LAL to release the fluorescent moiety 4-MU; is cleaved by GALNS to release the fluorescent moiety 4-MU; ex = 320 and 360 nm at low (1.97-6.72) and high (7.12-10.3) pH, respectively; em = 445-455 nm, increasing as pH decreases,
Catalog No. NC1480936