Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Methylamino)-3-nitrobenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. M242325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M242325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M242325G Supplier TCI America Supplier No. M242325G

Chemical Identifiers

CAS 41263-74-5
Molecular Formula C8H8N2O4
Molecular Weight (g/mol) 196.162
MDL Number MFCD00464322
InChI Key KSMLIIWEQBYUKA-UHFFFAOYSA-N
PubChem CID 283475
SMILES CNC1=C(C=C(C=C1)C(=O)O)[N+](=O)[O-]

Specifications

Melting Point 310°C
Color Yellow
Quantity 25 g
Formula Weight 196.16
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Methylamino)-3-nitrobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.