Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Methylamino)phenol Sulfate RS, Ricca Chemical
SDP

Catalog No. 48394 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
4839-4 120 mL Amber Glass Bottle
4839-16 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 4839-4 Supplier Ricca Chemical Company Supplier No. 48394
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5

Specifications

Grade ACS Reagent
Packaging Amber Glass Bottle
pH Acidic
Quantity 120 mL
Chemical Name or Material 4-(Methylamino)phenol Sulfate RS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.