Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Methylamino)pyridine, 99%

Catalog No. AAL1962614
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1962614 25 g
AAL1962603 1 g
AAL1962606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1962614 Supplier Thermo Scientific Chemicals Supplier No. L1962614
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 1121-58-0 | C6H8N2 | 108.14 g/mol

4-(Methylamino)pyridine is employed as efficient nucleophilic catalyst during the preparation of ultra-high surface area emulsion templated porous polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Methylamino)pyridine is employed as efficient nucleophilic catalyst during the preparation of ultra-high surface area emulsion templated porous polymers.

Solubility
Soluble in water, acetone, ethanol, ether and benzene.

Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1121-58-0
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00152625
InChI Key LSCYTCMNCWMCQE-UHFFFAOYSA-N
Synonym 4-methylamino pyridine, 4-methylaminopyridine, n-methyl-4-pyridinamine, 4-pyridinamine, n-methyl, pyridin-4-yl methyl amine, 4-mamp, dmap on polystyrene, pubchem12915, pyridin-4yl-methylamine, pyridin-4-yl-methylamine
PubChem CID 123098
IUPAC Name N-methylpyridin-4-amine
SMILES CNC1=CC=NC=C1

Specifications

Melting Point 124°C to 128°C
Quantity 25 g
Beilstein 108213
Solubility Information Soluble in water,acetone,ethanol,ether and benzene.
Formula Weight 108.14
Percent Purity 99%
Chemical Name or Material 4-(Methylamino)pyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.