Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylaminophenol sulfate, ACS reagent, 99.0-101.5%

Catalog No. AC434445000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC434445000 500 g Plastic bottle
AC434441000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC434445000 Supplier Thermo Scientific Chemicals Supplier No. 434445000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 55-55-0 | C14H18N2O2·H2SO4 | 344.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55-55-0
Molecular Formula C14H18N2O2·H2SO4
Molecular Weight (g/mol) 344.38
Synonym Metol

Specifications

Melting Point 260.0°C
Color White
Density 1.5770g/mL
Ignition Residue 0.1% max.
Packaging Plastic bottle
Linear Formula (CH3NHC6H4OH)2·H2SO4
Quantity 500 g
Merck Index 15,6091
Specific Gravity 1.577
Solubility Information Solubility in water: 50g/L (20°C). Other solubilities: soluble in 20 parts cold water,soluble in 6 parts boiling water,slightly soluble in alcohol,insoluble in ether
Formula Weight 344.38
Percent Purity 99.0-101.5%
Grade ACS Reagent
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylaminophenol sulfate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Very toxic to aquatic life with long lasting effects.
May cause an allergic skin reaction.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-237-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.