Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Methylanisole

Catalog No. 148091100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-809-1100ML 100 mL
14-809-11L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-809-1100ML Supplier Sigma Aldrich Supplier No. 148091100ML

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 4-methylanisole was used to prepare 5-methoxy-1,8-dimethyltetralin
  • It was also used as the solubilizate molecule to study the site of incorporation of solubilizates in sodium dodecyl sulfate (sds) micellar systems

Specifications

CAS 104-93-8
Density 0.969 g/mL (at 25°C (literature))
Boiling Point 174°C (lit.)
Molecular Formula C8H10O
Refractive Index n20/D 1.511 (literature)
Linear Formula CH3C6H4OCH3
Quantity 100 mL
MDL Number MFCD00008413
Molecular Weight (g/mol) 122.16
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.