Learn More
Medchemexpress LLC 4-methylbenzamide oxime | 19227-13-5 | 99.2% | 150.18 g/mol | C8H10N2O | 10 G
SDP
Catalog No. 5000210935
Encompass_Preferred
4-methylbenzamide oxime (CAS 19227-13-5) is a benzamidoxime derivative commonly used as a model substrate for mitochondrial amidoxime reducing component (mARC) catalytic enzyme reduction. It is supplied as a high-purity solid suitable for biochemical assays and research applications.
  • Model substrate for mARC enzyme reduction studies.
  • High purity suitable for biochemical and enzymatic assays.
  • Molecular formula C8H10N2O; molecular weight 150.18 g/mol.
  • Solid powder form with standard storage and handling requirements; consult SDS for safety information.
  • Available in multiple pack sizes, including a 10 G pack.
Catalog No. 50-002-10935 Supplier Medchemexpress LLC Supplier No. HYW05584510G
4-methylbenzamide oxime (CAS 19227-13-5) is a benzamidoxime derivative commonly used as a model substrate for mitochondrial amidoxime reducing component (mARC) catalytic enzyme reduction. It is supplied as a high-purity solid suitable for biochemical assays and research applications.
  • Model substrate for mARC enzyme reduction studies.
  • High purity suitable for biochemical and enzymatic assays.
  • Molecular formula C8H10N2O; molecular weight 150.18 g/mol.
  • Solid powder form with standard storage and handling requirements; consult SDS for safety information.
  • Available in multiple pack sizes, including a 10 G pack.