Learn More
Medchemexpress LLC 4-methylbenzamide oxime | 19227-13-5 | MFCD00019952 | 99.2% | 150.18 g/mol | C8H10N2O | 1 G
SDP
Catalog No. 5000210916
Encompass_Preferred
4-Methylbenzamide oxime is a small-molecule biochemical reagent used as a model benzamide oxime and as a substrate for mitochondrial amidoxime reducing component (mARC) enzyme assays. It is supplied as a high-purity solid for laboratory use.
  • High purity suitable for biochemical assays.
  • Functions as a substrate for mARC catalytic enzyme reduction.
  • Molecular formula C8H10N2O and molecular weight 150.18 g/mol.
  • Available in common laboratory pack sizes for small-scale experiments.
Catalog No. 50-002-10916 Supplier Medchemexpress LLC Supplier No. HYW0558451G
4-Methylbenzamide oxime is a small-molecule biochemical reagent used as a model benzamide oxime and as a substrate for mitochondrial amidoxime reducing component (mARC) enzyme assays. It is supplied as a high-purity solid for laboratory use.
  • High purity suitable for biochemical assays.
  • Functions as a substrate for mARC catalytic enzyme reduction.
  • Molecular formula C8H10N2O and molecular weight 150.18 g/mol.
  • Available in common laboratory pack sizes for small-scale experiments.