Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-methylbenzamide oxime | 19227-13-5 | MFCD00019952 | 99.2% | 150.18 g/mol | C8H10N2O | 25 G
SDP

Catalog No. 5000209154
Encompass_Preferred

4-Methylbenzamide oxime (CAS 19227-13-5) is a white to off-white solid used as a model benzamide oxime and as a substrate for mitochondrial amidoxime reducing component (mARC) enzyme reduction in biochemical assays. It is supplied at high purity and should be stored cold according to handling instructions in the SDS.

  • Model substrate for mARC enzyme assays
  • High purity (99.17%)
  • Molecular formula C8H10N2O; molecular weight 150.18 g/mol
  • White to off-white solid
  • Recommended storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years)

Catalog No. 50-002-09154 Supplier Medchemexpress LLC Supplier No. 25GHYW055845
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Methylbenzamide oxime (CAS 19227-13-5) is a white to off-white solid used as a model benzamide oxime and as a substrate for mitochondrial amidoxime reducing component (mARC) enzyme reduction in biochemical assays. It is supplied at high purity and should be stored cold according to handling instructions in the SDS.

  • Model substrate for mARC enzyme assays
  • High purity (99.17%)
  • Molecular formula C8H10N2O; molecular weight 150.18 g/mol
  • White to off-white solid
  • Recommended storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.