Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylbenzenesulphonylhydrazide, 97%

Catalog No. AAA1352922
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1352922 100 g
AAA1352914 25 g
AAA1352936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1352922 Supplier Thermo Scientific Chemicals Supplier No. A1352922
  • $91.30
    $77.65 / Each of 1

    Save $13.65

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1576-35-8 | C7H10N2O2S | 186.23 g/mol

p-Toluenesulfonyl hydrazide is used as a reagent for the preparation of tosylhydrazones. It was used to prepare dipyrazolo[1,5-a:4,3-c]pyridines and 1,2,3-selenadiazole derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Toluenesulfonyl hydrazide is used as a reagent for the preparation of tosylhydrazones. It was used to prepare dipyrazolo[1,5-a:4′,3′-c]pyridines and 1,2,3-selenadiazole derivatives.

Solubility
Soluble in water, alcohol.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1576-35-8
Molecular Formula C7H10N2O2S
Molecular Weight (g/mol) 186.23
MDL Number MFCD00007588
InChI Key ICGLPKIVTVWCFT-UHFFFAOYSA-N
Synonym 4-methylbenzenesulfonhydrazide, p-toluenesulfonhydrazide, tosylhydrazine, p-toluenesulfonyl hydrazide, tosylhydrazide, p-tosylhydrazine, p-toluenesulfonylhydrazine, celogen tsh, ptsh, 4-toluenesulfonyl hydrazide
PubChem CID 15303
IUPAC Name 4-methylbenzenesulfonohydrazide
SMILES CC1=CC=C(C=C1)S(=O)(=O)NN

Specifications

Melting Point 108°C to 111°C
Density 1.4
Quantity 100 g
UN Number UN3226
Beilstein 610130
Solubility Information Soluble in water,alcohol.
Formula Weight 186.23
Percent Purity 98%
Chemical Name or Material p-Toluenesulfonyl hydrazide
Hazard Category H242-H301-H319
Hazard Statement GHS H Statement
H242-H228-H301
Heating may cause a fire.
Flammable solid.
Toxic if swallowed.
Precautionary Statement P210-P220-P234-P264b-P270-P280-P301+P310-P305+P351+P338-P330-P370+P378q-P375-P380-P501c
DOTInformation Hazard Class: 4.1
EINECSNumber 216-407-3
RTECSNumber MW0210000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.