Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylbenzenethiol sodium salt, 98%, Thermo Scientific™

Catalog No. AAH5509506
Change view
Click to view available options
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AAH5509506 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5509506 Supplier Thermo Scientific Chemicals Supplier No. H5509506

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Methylbenzenethiol sodium salt is used as a pharmaceutical intermediate.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 10486-08-5
Molecular Formula C7H7NaS
Quantity 5g
MDL Number MFCD00066208
Synonym sodium toluene-p-thiolate, 4-methylbenzenethiol sodium salt, sodium 4-methylbenzenethiolate, sodium 4-methylphenyl sulfanide, sodium p-toluenethiolate, 4-methylbenzanethiol sodium, 4-methylphenyl sulfanyl sodium, 4-methylbenzenethiol, sodium salt
InChI Key PRORQUQEDGGDQO-UHFFFAOYSA-M
SMILES [Na+].CC1=CC=C([S-])C=C1
IUPAC Name sodium (4-methylphenyl)sulfanide
Molecular Weight (g/mol) 146.18
PubChem CID 5148259
Formula Weight 146.18
Percent Purity 98%
Chemical Name or Material 4-Methylbenzenethiol sodium salt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.