Learn More
Medchemexpress LLC 4-methylbenzothioamide | 2362-62-1 | 99.9% | 151.23 g/mol | C8H9NS | 10 G
SDP
Catalog No. 5000300634
Encompass_Preferred
4-Methylbenzothioamide is a thioamide building block used in medicinal chemistry and small-molecule synthesis. Supplied as an off-white to light-yellow solid with high assay purity, it is intended for use as a reagent or intermediate in drug discovery and organic synthesis applications.
  • High purity (99.87%) for reliable synthetic outcomes.
  • Off-white to light-yellow solid for straightforward handling and storage.
  • Supplied with a certificate of analysis confirming molecular composition and assay.
  • Storage conditions provided for both powder and solution states to preserve stability.
  • Useful building block for thioamide and heterocycle construction in medicinal chemistry.
Catalog No. 50-003-00634 Supplier Medchemexpress LLC Supplier No. HYW01739310G
Add to Cart
Add to Cart
4-Methylbenzothioamide is a thioamide building block used in medicinal chemistry and small-molecule synthesis. Supplied as an off-white to light-yellow solid with high assay purity, it is intended for use as a reagent or intermediate in drug discovery and organic synthesis applications.
  • High purity (99.87%) for reliable synthetic outcomes.
  • Off-white to light-yellow solid for straightforward handling and storage.
  • Supplied with a certificate of analysis confirming molecular composition and assay.
  • Storage conditions provided for both powder and solution states to preserve stability.
  • Useful building block for thioamide and heterocycle construction in medicinal chemistry.