Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylbenzotrifluoride 98.0+%, TCI America™
SDP

Catalog No. M29455G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M29455G 5 g
M294525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M29455G Supplier TCI America Supplier No. M29455G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6140-17-6
Molecular Formula C8H7F3
Molecular Weight (g/mol) 160.139
MDL Number MFCD00075476
InChI Key LRLRAYMYEXQKID-UHFFFAOYSA-N
Synonym 4-methylbenzotrifluoride, 1-methyl-4-trifluoromethyl benzene, 4-trifluoromethyl toluene, p-methylbenzotrifluoride, alpha,alpha,alpha-trifluoro-p-xylene, benzene, 1-methyl-4-trifluoromethyl, 4-methyl-1-trifluoromethyl benzene, 1-methyl-4-trifluormethyl benzol, pubchem1659, p-trifluoromethyltoluene
PubChem CID 80230
IUPAC Name 1-methyl-4-(trifluoromethyl)benzene
SMILES CC1=CC=C(C=C1)C(F)(F)F

Specifications

Color Colorless
Boiling Point 129°C
Quantity 5 g
UN Number 1993
Formula Weight 160.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methylbenzotrifluoride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.