Learn More
Cayman Chemical 4-Methylbenzylidene camphor, Purity: ≥98%, CAS: 36861-47-9, 5 g
SDP
Catalog No. 502955128
Encompass_Preferred
An ultraviolet light blocker used in cosmetics and sunscreen preparations that also has estrogenic activities, binding competitively to estrogen receptors and stimulating transactivation; changes reproductive organ and brain development in both males and females
Catalog No. 50-295-5128 Supplier Cayman Chemical Supplier No. 138405 G
An ultraviolet light blocker used in cosmetics and sunscreen preparations that also has estrogenic activities, binding competitively to estrogen receptors and stimulating transactivation; changes reproductive organ and brain development in both males and females