Learn More
Medchemexpress LLC 4-Methylbenzylidene camphor | 36861-47-9 | 99.87% | 254.37 | 250 MG
SDP
Catalog No. 5000384786
Encompass_Preferred
4-Methylbenzylidene camphor (4-MBC) is an endocrine disrupter that produces estrogen-like effects. It decreases the proliferation of human trophoblast cells and induces apoptosis. It also activates PI3K/AKT and ERK1/2 signaling pathways and elevates intracellular ROS production. 4-MBC functions as a ultraviolet (UV) filter and may hinder normal placental formation during early pregnancy. This product is for research use only and not intended for human use.
  • Endocrine disrupter with estrogen-like effects.
  • Decreases proliferation and induces apoptosis in human trophoblast cells.
  • Activates PI3K/AKT and ERK1/2 signaling pathways.
  • Elevates intracellular ROS production.
  • Functions as a ultraviolet (UV) filter.
Catalog No. 50-003-84786 Supplier Medchemexpress LLC Supplier No. 250MGHY17587R
4-Methylbenzylidene camphor (4-MBC) is an endocrine disrupter that produces estrogen-like effects. It decreases the proliferation of human trophoblast cells and induces apoptosis. It also activates PI3K/AKT and ERK1/2 signaling pathways and elevates intracellular ROS production. 4-MBC functions as a ultraviolet (UV) filter and may hinder normal placental formation during early pregnancy. This product is for research use only and not intended for human use.
  • Endocrine disrupter with estrogen-like effects.
  • Decreases proliferation and induces apoptosis in human trophoblast cells.
  • Activates PI3K/AKT and ERK1/2 signaling pathways.
  • Elevates intracellular ROS production.
  • Functions as a ultraviolet (UV) filter.