Learn More
Selleck Chemical LLC 4-Methylbenzylidene camphor S3704-25mg
SDP
Catalog No. 503110544
Encompass_Preferred
4-Methylbenzylidene camphor (Enzacamene 4-MBC) is an organic camphor derivative that is used in the cosmetic industry for its ability to protect the skin against UV specifically UV B radiation
Catalog No. 50-311-0544 Supplier Selleck Chemical LLC Supplier No. S370425MG
Add to Cart
Add to Cart
4-Methylbenzylidene camphor (Enzacamene 4-MBC) is an organic camphor derivative that is used in the cosmetic industry for its ability to protect the skin against UV specifically UV B radiation