Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Methylbiphenyl | 644-08-6 | MFCD00008544 | 100.0% | 168.23 g·mol^-1 | C13H12 | 25 G
SDP

Catalog No. 5000302318
Encompass_Preferred

4-Methylbiphenyl is an aromatic hydrocarbon and endogenous metabolite supplied as a high-purity analytical standard for research and analytical applications. It is available in solid and solution formats and is suitable as a reference material in biochemical and analytical studies.

  • High purity: 99.98%.
  • Molecular weight: 168.23 g·mol^-1.
  • Chemical formula: C13H12.
  • Appearance: white to off-white solid.
  • Soluble in DMSO at 100 mg/mL.
  • Available in multiple pack sizes, including 25 g solid and concentrated solutions.

Catalog No. 50-003-02318 Supplier Medchemexpress LLC Supplier No. 25GHYW017077
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Methylbiphenyl is an aromatic hydrocarbon and endogenous metabolite supplied as a high-purity analytical standard for research and analytical applications. It is available in solid and solution formats and is suitable as a reference material in biochemical and analytical studies.

  • High purity: 99.98%.
  • Molecular weight: 168.23 g·mol^-1.
  • Chemical formula: C13H12.
  • Appearance: white to off-white solid.
  • Soluble in DMSO at 100 mg/mL.
  • Available in multiple pack sizes, including 25 g solid and concentrated solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.