Learn More
Medchemexpress LLC 4-methylbiphenyl | 644-08-6 | MFCD00008544 | 100.0% | 168.23 g/mol | C13H12 | 1 ML
SDP
Catalog No. 5000300891
Encompass_Preferred
4-Methylbiphenyl is an endogenous metabolite supplied as a chemical standard for analytical and research applications. It is available as a ready-to-use 10 mM solution in DMSO and as a solid, suitable for HPLC, GC, and mass spectrometry workflows that require a reference standard.
  • High purity (99.98%) for accurate analytical results.
  • Ready-to-use 10 mM solution in DMSO reduces sample preparation time.
  • Compatible with HPLC, GC, and MS assays.
  • Available as solid in multiple sizes for method development and bulk use.
  • White to off-white solid appearance ensures batch consistency.
Catalog No. 50-003-00891 Supplier Medchemexpress LLC Supplier No. HYW01707710MM/1ML
4-Methylbiphenyl is an endogenous metabolite supplied as a chemical standard for analytical and research applications. It is available as a ready-to-use 10 mM solution in DMSO and as a solid, suitable for HPLC, GC, and mass spectrometry workflows that require a reference standard.
  • High purity (99.98%) for accurate analytical results.
  • Ready-to-use 10 mM solution in DMSO reduces sample preparation time.
  • Compatible with HPLC, GC, and MS assays.
  • Available as solid in multiple sizes for method development and bulk use.
  • White to off-white solid appearance ensures batch consistency.