Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-methylbiphenyl | 644-08-6 | MFCD00008544 | 100.0% | 168.23 g·mol⁻¹ | C13H12 | 10 G
SDP

Catalog No. 5000300335
Encompass_Preferred

4-Methylbiphenyl is an aromatic hydrocarbon used as a laboratory research chemical and reference standard. It is described as an endogenous metabolite and is supplied as a high-purity solid for analytical and synthetic applications. Consult the product SDS and COA for handling, storage, and safety information.

  • Aromatic hydrocarbon and endogenous metabolite.
  • High purity suitable for analytical use.
  • Solid, white to off-white appearance.
  • Molecular formula C13H12; molecular weight 168.23 g·mol⁻¹.
  • Soluble in common organic solvents such as DMSO.
  • Intended for research use; follow SDS guidance for safe handling.

Catalog No. 50-003-00335 Supplier Medchemexpress LLC Supplier No. HYW01707710G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Methylbiphenyl is an aromatic hydrocarbon used as a laboratory research chemical and reference standard. It is described as an endogenous metabolite and is supplied as a high-purity solid for analytical and synthetic applications. Consult the product SDS and COA for handling, storage, and safety information.

  • Aromatic hydrocarbon and endogenous metabolite.
  • High purity suitable for analytical use.
  • Solid, white to off-white appearance.
  • Molecular formula C13H12; molecular weight 168.23 g·mol⁻¹.
  • Soluble in common organic solvents such as DMSO.
  • Intended for research use; follow SDS guidance for safe handling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.