Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-methylbiphenyl | 644-08-6 | MFCD00008544 | 100.0% | 168.23 g/mol | C13H12 | 5 G
SDP

Catalog No. 5000299859
Encompass_Preferred

4-Methylbiphenyl is a biphenyl derivative and endogenous metabolite provided as a white to off-white solid for research applications. It is supplied with high purity and supporting documentation for analytical and chemical biology use.

  • High purity (99.98%) suitable for analytical workflows.
  • Chemical formula C13H12 and molecular weight 168.23 g/mol.
  • Supplied as a white to off-white solid for easy handling and storage.
  • Soluble in DMSO for preparation of stock solutions.
  • Certificate of analysis and safety data sheet available.
  • Multiple pack sizes available to fit experimental needs.

Catalog No. 50-002-99859 Supplier Medchemexpress LLC Supplier No. HYW0170775G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Methylbiphenyl is a biphenyl derivative and endogenous metabolite provided as a white to off-white solid for research applications. It is supplied with high purity and supporting documentation for analytical and chemical biology use.

  • High purity (99.98%) suitable for analytical workflows.
  • Chemical formula C13H12 and molecular weight 168.23 g/mol.
  • Supplied as a white to off-white solid for easy handling and storage.
  • Soluble in DMSO for preparation of stock solutions.
  • Certificate of analysis and safety data sheet available.
  • Multiple pack sizes available to fit experimental needs.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.