Learn More
Medchemexpress LLC 4-methylbiphenyl | 644-08-6 | MFCD00008544 | 100.0% | 168.23 g/mol | C13H12 | 50 G
SDP
Catalog No. 5000299487
Encompass_Preferred
4-Methylbiphenyl is an aromatic hydrocarbon and endogenous metabolite supplied as a high-purity solid for research and analytical use. It is commonly used as an analytical reference, in metabolomics studies, and as a synthetic building block in organic chemistry.
  • High purity suitable for analytical applications.
  • Solid white to off-white physical form.
  • Soluble in common organic solvents such as alcohol and ether.
  • Useful as an analytical standard and metabolite reference.
  • Stable under normal laboratory storage conditions.
Catalog No. 50-002-99487 Supplier Medchemexpress LLC Supplier No. HYW01707750G
4-Methylbiphenyl is an aromatic hydrocarbon and endogenous metabolite supplied as a high-purity solid for research and analytical use. It is commonly used as an analytical reference, in metabolomics studies, and as a synthetic building block in organic chemistry.
  • High purity suitable for analytical applications.
  • Solid white to off-white physical form.
  • Soluble in common organic solvents such as alcohol and ether.
  • Useful as an analytical standard and metabolite reference.
  • Stable under normal laboratory storage conditions.