Learn More
Medchemexpress LLC HY-W012814 100mg Medchemexpress, 4-Methylcatechol CAS:452-86-8 Purity:>98%
SDP
Catalog No. 501970692
Encompass_Preferred
Medchemexpress, HY-W012814 100mg 4-Methylcatechol CAS:452-86-8 4-Methylcatechol, a metabolite of p-toluate, is a substrate as well as a suicide inhibitor of Catechol 2,3-Dioxygenase. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0692 Supplier Medchemexpress LLC Supplier No. HYW012814100MG
Medchemexpress, HY-W012814 100mg 4-Methylcatechol CAS:452-86-8 4-Methylcatechol, a metabolite of p-toluate, is a substrate as well as a suicide inhibitor of Catechol 2,3-Dioxygenase. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.