Learn More
Apexbio Technology LLC 4-Methylcatechol 452-86-8 100mg
SDP
Catalog No. 5000094639
Encompass_Preferred
4-Methylcatechol (CAS 452-86-8) is a synthetic organic molecule structurally characterized by a methyl group substituted on the benzene ring of a 1 2-benzenediol framework classifying it within the catechol family This compound is known to exert biological effects primarily through modulation of oxidative stress pathways notably as a substrate and regulator of catechol-O-methyltransferase (COMT) and related metabolic enzymes 4-Methylcatechol demonstrates activity in modulating cellular redox states and has been studied for its capacity to influence neuronal signaling and protective mechanisms in neurobiology It is commonly used in experimental models investigating oxidative metabolism neuroprotection and mechanisms of catecholamine metabolism
Catalog No. 50-000-94639 Supplier Apexbio Technology LLC Supplier No. C7139100
4-Methylcatechol (CAS 452-86-8) is a synthetic organic molecule structurally characterized by a methyl group substituted on the benzene ring of a 1 2-benzenediol framework classifying it within the catechol family This compound is known to exert biological effects primarily through modulation of oxidative stress pathways notably as a substrate and regulator of catechol-O-methyltransferase (COMT) and related metabolic enzymes 4-Methylcatechol demonstrates activity in modulating cellular redox states and has been studied for its capacity to influence neuronal signaling and protective mechanisms in neurobiology It is commonly used in experimental models investigating oxidative metabolism neuroprotection and mechanisms of catecholamine metabolism