Learn More
Medchemexpress LLC 4-Methylcatechol | 452-86-8 | MFCD00002205 | 99.5% | 124.14 | 1 ML
SDP
Catalog No. 5000454922
Encompass_Preferred
4-Methylcatechol is an intermediate involved in the degradation of certain alkylbenzenes and acts as an orally active suicide inhibitor of catechol 2,3-dioxygenase. It has been shown to induce apoptosis in melanoma cells through oxidative stress. While it exhibits antiplatelet and blood pressure-lowering activities, some studies suggest it may have carcinogenic properties.
  • Induces cell cycle arrest and apoptosis via the mitochondrial pathway.
  • Inhibits protein oxidation in beef.
  • Associated with vasodilatory and blood pressure-lowering effects in animal models.
  • Has shown carcinogenic effects in forestomach and glandular stomach in rat studies.
Catalog No. 50-004-54922 Supplier Medchemexpress LLC Supplier No. HYW01281410MM/1ML
4-Methylcatechol is an intermediate involved in the degradation of certain alkylbenzenes and acts as an orally active suicide inhibitor of catechol 2,3-dioxygenase. It has been shown to induce apoptosis in melanoma cells through oxidative stress. While it exhibits antiplatelet and blood pressure-lowering activities, some studies suggest it may have carcinogenic properties.
  • Induces cell cycle arrest and apoptosis via the mitochondrial pathway.
  • Inhibits protein oxidation in beef.
  • Associated with vasodilatory and blood pressure-lowering effects in animal models.
  • Has shown carcinogenic effects in forestomach and glandular stomach in rat studies.