Learn More
Medchemexpress LLC 4-Methylcatechol-d3 | 83719-41-9 | 98.4% | 127.16 | 5 MG
SDP
Catalog No. 5000454799
Encompass_Preferred
4-Methylcatechol-d3 is the deuterium labeled 4-Methylcatechol. 4-Methylcatechol, a metabolite of p-toluate, is a substrate as well as a suicide inhibitor of Catechol 2,3-Dioxygenase.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-54799 Supplier Medchemexpress LLC Supplier No. HYW012814S5MG
4-Methylcatechol-d3 is the deuterium labeled 4-Methylcatechol. 4-Methylcatechol, a metabolite of p-toluate, is a substrate as well as a suicide inhibitor of Catechol 2,3-Dioxygenase.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.