Learn More
Medchemexpress LLC 4α-Methylcholesterol | 15073-00-4 | 5 MG
SDP
Catalog No. 5000419256
Encompass_Preferred
4α-Methylcholesterol is a cholesterol derivative that can oxidize 3-hydroxy steroid, with an apparent Km of 12.6 μM.
  • It is a cholesterol derivative.
  • Can oxidize 3-hydroxy steroid.
  • Has a purity of 99.90%.
  • Appears as a solid.
  • Color is white to off-white.
Catalog No. 50-004-19256 Supplier Medchemexpress LLC Supplier No. HYN85315MG
4α-Methylcholesterol is a cholesterol derivative that can oxidize 3-hydroxy steroid, with an apparent Km of 12.6 μM.
  • It is a cholesterol derivative.
  • Can oxidize 3-hydroxy steroid.
  • Has a purity of 99.90%.
  • Appears as a solid.
  • Color is white to off-white.