Learn More
Medchemexpress LLC HY-112734 10mg Medchemexpress, 4'-Methylchrysoeriol CAS:4712-12-3 Purity:>98%
SDP
Catalog No. 501969162
Encompass_Preferred
Medchemexpress, HY-112734 10mg 4'-Methylchrysoeriol CAS:4712-12-3 4'-Methylchrysoeriol is a potent inhibitor of Cytochrome P450 enzymes, with an IC50 of 19 nM for human P450 1B1-dependent EROD. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9162 Supplier Medchemexpress LLC Supplier No. HY11273410MG
Medchemexpress, HY-112734 10mg 4'-Methylchrysoeriol CAS:4712-12-3 4'-Methylchrysoeriol is a potent inhibitor of Cytochrome P450 enzymes, with an IC50 of 19 nM for human P450 1B1-dependent EROD. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.