Learn More
Medchemexpress LLC 4'-Methylchrysoeriol | 4712-12-3 | 99.2% | 314.29 | 1 ML
SDP
Catalog No. 5000437927
Encompass_Preferred
4'-Methylchrysoeriol is a potent inhibitor of Cytochrome P450 enzymes, with an IC50 of 19 nM for human P450 1B1-dependent EROD. This chemical is designed for research use, offering high purity for reliable experimental results.
  • Potent inhibitor of Cytochrome P450 enzymes.
  • Soluble in DMSO.
  • Can be used for in vivo experiments.
  • Long-term stability as powder at -20°C.
  • Stable in solvent at -80°C for up to 6 months.
Catalog No. 50-004-37927 Supplier Medchemexpress LLC Supplier No. HY11273410MM/1ML
Add to Cart
Add to Cart
4'-Methylchrysoeriol is a potent inhibitor of Cytochrome P450 enzymes, with an IC50 of 19 nM for human P450 1B1-dependent EROD. This chemical is designed for research use, offering high purity for reliable experimental results.
  • Potent inhibitor of Cytochrome P450 enzymes.
  • Soluble in DMSO.
  • Can be used for in vivo experiments.
  • Long-term stability as powder at -20°C.
  • Stable in solvent at -80°C for up to 6 months.