Learn More
Medchemexpress LLC 4-methylcinnamic acid | 1866-39-3 | MFCD00002697 | 99.0% | 162.19 g/mol | C10H10O2 | 1 G
SDP
Catalog No. 5000302786
Encompass_Preferred
4-Methylcinnamic acid is a cinnamic acid derivative reported to have antifungal activity and is used in research to study antifungal effects and structure-activity relationships among cinnamic acid derivatives. It is a small organic acid (C10H10O2, MW 162.19 g/mol) typically supplied in research pack sizes for in vitro studies.
  • Cinnamic acid derivative with reported antifungal activity.
  • Suitable for in vitro antifungal and structure-activity studies.
  • Chemical formula C10H10O2, molecular weight 162.19 g/mol.
  • Predominantly trans isomer reported; high purity (~99%) available.
  • Provided in small research pack sizes for laboratory use.
Catalog No. 50-003-02786 Supplier Medchemexpress LLC Supplier No. HYW0153991G
4-Methylcinnamic acid is a cinnamic acid derivative reported to have antifungal activity and is used in research to study antifungal effects and structure-activity relationships among cinnamic acid derivatives. It is a small organic acid (C10H10O2, MW 162.19 g/mol) typically supplied in research pack sizes for in vitro studies.
  • Cinnamic acid derivative with reported antifungal activity.
  • Suitable for in vitro antifungal and structure-activity studies.
  • Chemical formula C10H10O2, molecular weight 162.19 g/mol.
  • Predominantly trans isomer reported; high purity (~99%) available.
  • Provided in small research pack sizes for laboratory use.