Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-methylcinnamic acid | 1866-39-3 | MFCD00002697 | 99.6% | 162.19 | C10H10O2 | 1 ML
SDP

Catalog No. 5000303266
Encompass_Preferred

4-methylcinnamic acid is a cinnamic acid derivative used in research with reported antifungal activity. It is supplied for in vitro and in vivo studies and is provided as powder or concentrated stock solutions with guidance for formulation and storage.

  • Cas number 1866-39-3.
  • Molecular formula C10H10O2.
  • Molecular weight 162.19 g/mol.
  • Purity approximately 99.6%.
  • Available as powder and in-solvent preparations.
  • Solvent guidance: DMSO for high-concentration stocks.
  • Stock preparation guidance for 1, 5, and 10 mM solutions from mg quantities.
  • Storage recommendations: powder and in-solution temperature ranges with multi-year stability.

Catalog No. 50-003-03266 Supplier Medchemexpress LLC Supplier No. HYW01539910MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-methylcinnamic acid is a cinnamic acid derivative used in research with reported antifungal activity. It is supplied for in vitro and in vivo studies and is provided as powder or concentrated stock solutions with guidance for formulation and storage.

  • Cas number 1866-39-3.
  • Molecular formula C10H10O2.
  • Molecular weight 162.19 g/mol.
  • Purity approximately 99.6%.
  • Available as powder and in-solvent preparations.
  • Solvent guidance: DMSO for high-concentration stocks.
  • Stock preparation guidance for 1, 5, and 10 mM solutions from mg quantities.
  • Storage recommendations: powder and in-solution temperature ranges with multi-year stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.