Learn More
Medchemexpress LLC HY-W015399 500mg Medchemexpress, 4-Methylcinnamic acid CAS:1866-39-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW015399500MG
Encompass_Preferred
Medchemexpress, HY-W015399 500mg 4-Methylcinnamic acid CAS:1866-39-3 4-Methylcinnamic acid, a Cinnamic acid analog, can be used as a intervention catalyst for overcoming antifungal tolerance. 4-Methylcinnamic acid can improve the potency of cell wall-disrupting agents. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3375