Learn More
Medchemexpress LLC HY-N4288 100mg Medchemexpress, 4-Methylesculetin CAS:529-84-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN4288100MG
Encompass_Preferred
Medchemexpress, HY-N4288 100mg 4-Methylesculetin CAS:529-84-0 4-Methylesculetin is an orally active natural coumarin derivative, with potent anti-oxidant and anti-inflammatory activities. 4-Methylesculetin inhibits myeloperoxidase activity and reduces IL-6 level [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3951