Learn More
Cayman Chemical 4-Methylethcathinone metabolite (hydrochloride) ((±)-Ephedrine stereochemistry), Purity: ≥98%, 1 mg
SDP
Catalog No. 502969984
Encompass_Preferred
A metabolite of 4-MEC that features conversion of the β-keto group to β-hydroxy and is an enantiomeric mixture of the R,S and S,R orientations at carbons one and two, as in ephedrine; intended for research and forensic applications
Catalog No. 50-296-9984 Supplier Cayman Chemical Supplier No. 147581 MG
A metabolite of 4-MEC that features conversion of the β-keto group to β-hydroxy and is an enantiomeric mixture of the R,S and S,R orientations at carbons one and two, as in ephedrine; intended for research and forensic applications