Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylheptane 98.0+%, TCI America™
SDP

Catalog No. M05201ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M05201ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M05201ML Supplier TCI America Supplier No. M05201ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 589-53-7
Molecular Formula C8H18
Molecular Weight (g/mol) 114.23
MDL Number MFCD00009409
InChI Key CHBAWFGIXDBEBT-UHFFFAOYSA-N
PubChem CID 11512
ChEBI CHEBI:67193
IUPAC Name 4-methylheptane
SMILES CCCC(C)CCC

Specifications

Color Colorless
Boiling Point 117°C
Quantity 1 mL
UN Number 1262
Formula Weight 114.23
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methylheptane
EINECSNumber (2)-0008
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.