Learn More
Apexbio Technology LLC 4-Methylhistamine dihydrochloride 36376-47-3 5mg
SDP
Catalog No. 5000087817
Encompass_Preferred
4-Methylhistamine dihydrochloride (CAS 36376-47-3) is a selective agonist for the histamine H4 receptor (H4R) displaying a pEC50 of 7 4 0 1 at human H4R It exhibits over 100-fold selectivity for H4R relative to H3R and H2R and over 100 000-fold compared to H1R While affinity for human H4R is moderate it binds rat and mouse H4R with higher affinity (Ki values of 73 nM and 55 nM respectively) In in vivo models 4-Methylhistamine dihydrochloride modulates inflammatory signaling elevating TNF- and NF- B expression via JAK/STAT pathways in response to LPS-induced joint inflammation It serves as a valuable tool in studies of H4R-mediated immune regulation and inflammatory mechanisms
Catalog No. 50-000-87817 Supplier Apexbio Technology LLC Supplier No. B70265
4-Methylhistamine dihydrochloride (CAS 36376-47-3) is a selective agonist for the histamine H4 receptor (H4R) displaying a pEC50 of 7 4 0 1 at human H4R It exhibits over 100-fold selectivity for H4R relative to H3R and H2R and over 100 000-fold compared to H1R While affinity for human H4R is moderate it binds rat and mouse H4R with higher affinity (Ki values of 73 nM and 55 nM respectively) In in vivo models 4-Methylhistamine dihydrochloride modulates inflammatory signaling elevating TNF- and NF- B expression via JAK/STAT pathways in response to LPS-induced joint inflammation It serves as a valuable tool in studies of H4R-mediated immune regulation and inflammatory mechanisms