Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylimidazole, 98%, ACROS Organics™

Click to view available options
Quantity:
50g
250g
Packaging:
Glass bottle
822-36-6
C4H6N2
82.11
MFCD00005201
XLSZMDLNRCVEIJ-UHFFFAOYSA-N
4-methylimidazole,4-methyl-1h-imidazole,4 5-methylimidazole,1h-imidazole, 4-methyl,4-me-i,5-methylimidazole,imidazole, 4-methyl,4 or 5-methylimidazole,4-mei,4-methylimidazol
13195
CHEBI:40035
5-methyl-1H-imidazole
CC1=CN=CN1
822-36-6
C4H6N2
82.11
MFCD00005201
XLSZMDLNRCVEIJ-UHFFFAOYSA-N
4-methylimidazole,4-methyl-1h-imidazole,4 5-methylimidazole,1h-imidazole, 4-methyl,4-me-i,5-methylimidazole,imidazole, 4-methyl,4 or 5-methylimidazole,4-mei,4-methylimidazol
13195
CHEBI:40035
5-methyl-1H-imidazole
CC1=CN=CN1
Melting Point 52.0°C to 56.0°C
Water 0.5% max. (K.F.)
Infrared Spectrum Authentic
Packaging Glass bottle
Color Brown to Brown-Red
Flash Point 157°C
Boiling Point 263.0°C
Quantity 50g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Beilstein 23, 69
Solubility Information Solubility in water: soluble. Other solubilities: soluble in almost all common organic solvents
Formula Weight 82.11
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 4-Methylimidazole, 98%
Show More Show Less

GHS H Statement
Toxic in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.

GHS Signal Word: Danger

EINECSNumber : 212-497-3

RTECSNumber : NI7350000

TSCA : TSCA