Learn More
Medchemexpress LLC 4-Methylimidazole-d6 | 1219804-79-1 | 98.3% | 88.14 | 25 MG
SDP
Catalog No. 5000452661
Encompass_Preferred
4-Methylimidazole-d6 is the deuterium labeled 4-Methylimidazole.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-52661 Supplier Medchemexpress LLC Supplier No. 25MGHYW012814S2
4-Methylimidazole-d6 is the deuterium labeled 4-Methylimidazole.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.