Learn More
Medchemexpress LLC 4-Methylimidazole-d6 | 1219804-79-1 | 98.3% | 88.14 | 5 MG
SDP
Catalog No. 5000454800
Encompass_Preferred
4-Methylimidazole-d6 is a deuterium labeled compound primarily used for research. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes have gained attention due to their potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Features stable heavy isotopes.
Catalog No. 50-004-54800 Supplier Medchemexpress LLC Supplier No. HYW012814S25MG
4-Methylimidazole-d6 is a deuterium labeled compound primarily used for research. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes have gained attention due to their potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Features stable heavy isotopes.