Learn More
Medchemexpress LLC 4-Methylimidazole-d6 | 1219804-79-1 | MFC4D6N2 | 98.3% | 88.14 | 10 MG
SDP
Catalog No. 5000454967
Encompass_Preferred
This is a stable isotope-labeled compound. It can be utilized as a tracer and as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS. The incorporation of stable heavy isotopes like deuterium into drug molecules is primarily for quantitation during the drug development process, and deuteration can influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotope.
  • Potential to affect pharmacokinetic and metabolic profiles.
Catalog No. 50-004-54967 Supplier Medchemexpress LLC Supplier No. HYW012814S210MG
This is a stable isotope-labeled compound. It can be utilized as a tracer and as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS. The incorporation of stable heavy isotopes like deuterium into drug molecules is primarily for quantitation during the drug development process, and deuteration can influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotope.
  • Potential to affect pharmacokinetic and metabolic profiles.