Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylimidazole 98.0+%, TCI America™
SDP

Catalog No. M0636100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0636100G 100 g
M063625G 25 g
M0636500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M0636100G Supplier TCI America Supplier No. M0636100G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 822-36-6
Molecular Formula C4H6N2
Molecular Weight (g/mol) 82.11
MDL Number MFCD00005201
InChI Key XLSZMDLNRCVEIJ-UHFFFAOYSA-N
Synonym 4-methylimidazole, 4-methyl-1h-imidazole, 4 5-methylimidazole, 1h-imidazole, 4-methyl, 4-me-i, 5-methylimidazole, imidazole, 4-methyl, 4 or 5-methylimidazole, 4-mei, 4-methylimidazol
PubChem CID 13195
ChEBI CHEBI:40035
IUPAC Name 5-methyl-1H-imidazole
SMILES CC1=CN=CN1

Specifications

Melting Point 52°C
Color Yellow
Boiling Point 263°C
Quantity 100 g
UN Number 3263
Formula Weight 82.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylimidazole
EINECSNumber (5)-5709
RTECSNumber NI7350000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.