Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-methylisoxazole-5-carboxylic acid

Catalog No. CDS012084 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS012084 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS012084 Supplier Sigma Aldrich Supplier No. CDS012084100MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Methyl 3-(methylthio)propionate is the major ester present in the flavour profile of juices made from fresh-cut pineapple fruits.methyl 3-(methylthio)propionate was used to prepare 3-methiolpropionate(mmpa) to study the demethylation of dimethylsulfoniopropionate and mmpa to 3-mercaptopropionate by aerobic marine bacterium
  • Methyl 3-(methylthio)propionate was used to prepare 3-(methylthio)propionic acid to examine the methionine catabolism by lactococci by 13c nuclear magnetic resonance and gas chromatography

Specifications

Molecular Formula C5H5NO3
Linear Formula C5H5NO3
MDL Number MFCD08235209
Quantity 25 g
Molecular Weight (g/mol) 127.1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.