Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Methylnitrosamino)-1-(3-pyridyl)-1-butanol, ≥92% (TLC), MilliporeSigma™ Supelco™

Catalog No. 111015446 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5446 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5446 Supplier MilliporeSigma Supelco Supplier No. 5977310MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Standard for Supelco MIP SPE cartridges. For more information request Supelco Literature T407075, T706031

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 76014-81-8
Physical Form Neat
Linear Formula C10H15N3O2
MDL Number MFCD00870462
Synonym NNAL
Type Analytical Standard
Quantity 10 mg
Formula Weight 209.25
Percent Purity ≥92% (TLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.