Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Methyloctanoic acid | 54947-74-9 | 99.85% | 158.24 | 100 G
SDP

Catalog No. 5000453629
Encompass_Preferred

4-Methyloctanoic acid is a natural compound primarily responsible for the characteristic "goaty" or "sheepy" flavor found in the milk of sheep and goats. It is intended for research use only and is not sold to patients.

  • Purity: 99.85%
  • Appearance: Liquid (density: 0.91 g/cm³)
  • Color: Colorless to light yellow
  • Initial source: Animals (sheep milk)
  • Storage: Store at room temperature for 3 years; in solvent: -80°C for 2 years, -20°C for 1 year.
  • Solubility (in vitro): 100 mg/mL in DMSO (631.95 mM); requires ultrasonic and freshly opened DMSO due to hygroscopic nature.

Catalog No. 50-004-53629 Supplier Medchemexpress LLC Supplier No. HYW010178100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Methyloctanoic acid is a natural compound primarily responsible for the characteristic "goaty" or "sheepy" flavor found in the milk of sheep and goats. It is intended for research use only and is not sold to patients.

  • Purity: 99.85%
  • Appearance: Liquid (density: 0.91 g/cm³)
  • Color: Colorless to light yellow
  • Initial source: Animals (sheep milk)
  • Storage: Store at room temperature for 3 years; in solvent: -80°C for 2 years, -20°C for 1 year.
  • Solubility (in vitro): 100 mg/mL in DMSO (631.95 mM); requires ultrasonic and freshly opened DMSO due to hygroscopic nature.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.