Learn More
Sigma Aldrich Fine Chemicals Biosciences 4 Methyloctanoic acid100G
Catalog No. 502903138
Encompass
4-Methyloctanoic acid is a branched fatty acid commonly used in the flavor and fragrance industries. This compound gives sweaty and goaty flavors to the mutton meat and goat milk cheese respectively.
Catalog No. 50-290-3138 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W357502100GK
4-Methyloctanoic acid is a branched fatty acid commonly used in the flavor and fragrance industries. This compound gives sweaty and goaty flavors to the mutton meat and goat milk cheese respectively.