Learn More
Cayman Chemical 4-Methylpentanoic Acid-d12, Purity: ≥99% deuterated forms (d1-d12), CAS: 116287-57-1, 50 mg
SDP
Catalog No. 502940236
Encompass_Preferred
An internal standard for the quantification of 4-methylpentanoic acid by GC- or LC-MS
Catalog No. 50-294-0236 Supplier Cayman Chemical Supplier No. 2787350 MG
An internal standard for the quantification of 4-methylpentanoic acid by GC- or LC-MS