Learn More
Medchemexpress LLC 4-methylphenyl 1-thio-β-D-glucopyranoside | 1152-39-2 | MFCD11045365 | >98.0% | 286.34 g/mol | C13H18O5S | 1 G
SDP
Catalog No. 5000285105
Encompass_Preferred
4-Methylphenyl 1-thio-β-D-glucopyranoside is a thioglycoside reagent used in glycobiology and carbohydrate research. It is provided as a solid research reagent for laboratory use and is characterized by standard chemical identifiers for unambiguous sourcing and handling information.
  • Used as a thioglycoside reagent in glycobiology and carbohydrate research.
  • Suitable for glycan synthesis and enzymatic assays.
  • Reported high purity (≥98%) from multiple suppliers.
  • Available in multiple packaging sizes, including 1 G.
  • Documented identifiers: CAS 1152-39-2, MFCD11045365.
Catalog No. 50-002-85105 Supplier Medchemexpress LLC Supplier No. HYW3571251G
4-Methylphenyl 1-thio-β-D-glucopyranoside is a thioglycoside reagent used in glycobiology and carbohydrate research. It is provided as a solid research reagent for laboratory use and is characterized by standard chemical identifiers for unambiguous sourcing and handling information.
  • Used as a thioglycoside reagent in glycobiology and carbohydrate research.
  • Suitable for glycan synthesis and enzymatic assays.
  • Reported high purity (≥98%) from multiple suppliers.
  • Available in multiple packaging sizes, including 1 G.
  • Documented identifiers: CAS 1152-39-2, MFCD11045365.